Thermo Scientific Chemicals

Potassium phosphate, 0.5M buffer soln., pH 7.4

Catalog number: J61413.AK
250 mL, Each
Thermo Scientific Chemicals

Potassium phosphate, 0.5M buffer soln., pH 7.4

Catalog number: J61413.AK
250 mL, Each
Quantity
Catalog number: J61413.AK
also known as J61413-AK
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7778-77-0
IUPAC Name
potassium dihydrogen phosphate
Molecular Formula
H2KO4P
InChI Key
GNSKLFRGEWLPPA-UHFFFAOYSA-M
SMILES
[K+].OP(O)([O-])=O
Appearance (Color)
Clear colorless
Form
Liquid
Comment
Filtered through 0.22 micron filter.
pH
7.40 ± 0.20

Description

Potassium phosphate is used as a buffer for the determination of pH. It is also used in pharmaceutical production, urinary acidifier, paper processing, nutrient solutions and yeast foods.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Potassium phosphate is used as a buffer for the determination of pH. It is also used in pharmaceutical production, urinary acidifier, paper processing, nutrient solutions and yeast foods.

Solubility
Miscible with water. Immiscible with alcohol.

Notes
Hygroscopic. Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text