Thermo Scientific Chemicals

Guanosine-5'-triphosphate disodium salt

Catalog number: J61414.ME
500 mg, Each
Thermo Scientific Chemicals

Guanosine-5'-triphosphate disodium salt

Catalog number: J61414.ME
500 mg, Each
Quantity
Catalog number: J61414.ME
also known as J61414-ME
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
56001-37-7
IUPAC Name
disodium phosphono ({[(2R,3S,4R,5R)-5-(2-amino-6-oxo-6,9-dihydro-1H-purin-9-yl)-3,4-dihydroxyoxolan-2-yl]methyl phosphonato}oxy)phosphonate
Molecular Formula
C10H14N5Na2O14P3
InChI Key
FIZIYLKEXVIRHJ-KHRSEZDTNA-L
SMILES
[Na+].[Na+].NC1=NC2=C(N=CN2[C@@H]2O[C@H](COP([O-])(=O)OP([O-])(=O)OP(O)(O)=O)[C@@H](O)[C@H]2O)C(=O)N1
Assay (HPLC)
90.0% min.
Loss on Drying
15.0% max.
Appearance (Color)
White to off-white
Form
Powder

Description

Guanosine 5?-triphosphate (GTP), a purine trinucleotide, is a substrate for RNA polymerases and a wide range of GTPase including G-protein coupled receptor (GPCR) GTPases and cell signaling and cycling associated guanine nucleotide exhange factors (GEF). Guanosine 5?-Triphosphate, Disodium Salt is a phosphoryl donor in protein synthesis and in signal transduction.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Guanosine 5′-triphosphate (GTP), a purine trinucleotide, is a substrate for RNA polymerases and a wide range of GTPase including G-protein coupled receptor (GPCR) GTPases and cell signaling and cycling associated guanine nucleotide exhange factors (GEF). Guanosine 5′-Triphosphate, Disodium Salt is a phosphoryl donor in protein synthesis and in signal transduction.

Solubility
Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature: 2 - 8°C. Keep away from strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text