Thermo Scientific Chemicals

Tween 20 washing buffer, 2% in PBS (20X)

Catalog number: J61419.K3
2 L, Each
Thermo Scientific Chemicals

Tween 20 washing buffer, 2% in PBS (20X)

Catalog number: J61419.K3
2 L, Each
Quantity
Catalog number: J61419.K3
also known as J61419-K3
Price (USD)
Quantity
-
Have Questions?
Composition
Potassium Phosphate Monobasic: 35.2 mM
Composition
Sodium Phosphate Dibasic: 202 mM
Form
Liquid
pH
7.25-7.55
Composition
Potassium Chloride: 54 mM

Description

PBS (Phosphate Buffered Saline) and Tween 20 are used as a wash buffer in immunolabeling techniques. Tween 20 is added to promote more effective washings, resulting in decreased non-specific background staining. Tween 20 is also known to promote reagent spreading in automated staining methodologies. 20X PBS Tween-20 is a space-saving stock solution that is ideal for preparing PBS-Tween (PBS-T) wash buffers for ELISA, Western and other immunoassays as well as a blocking buffer for plate-based assays.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text