Telmisartan, 98%, Thermo Scientific Chemicals
Telmisartan, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Telmisartan, 98%, Thermo Scientific Chemicals

An angiotensin II receptor antagonist
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number J61441.MD
also known as J61441-MD
Price (USD)/ Each
73.10
-
Add to cart
Quantity:
250 mg
Request bulk or custom format
Price (USD)/ Each
73.10
Add to cart
Telmisartan, 98%, Thermo Scientific Chemicals
Catalog numberJ61441.MD
Price (USD)/ Each
73.10
-
Add to cart
Chemical Identifiers
CAS144701-48-4
IUPAC Name4'-{[4-methyl-6-(1-methyl-1H-1,3-benzodiazol-2-yl)-2-propyl-1H-1,3-benzodiazol-1-yl]methyl}-[1,1'-biphenyl]-2-carboxylic acid
Molecular FormulaC33H30N4O2
InChI KeyRMMXLENWKUUMAY-UHFFFAOYSA-N
SMILESCCCC1=NC2=C(C)C=C(C=C2N1CC1=CC=C(C=C1)C1=CC=CC=C1C(O)=O)C1=NC2=CC=CC=C2N1C
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystalline powder or powder or crystals
Appearance (Color)White to off-white
Assay from Suppliers CofA≥97.5%

Telmisartan also acts as a partial agonist of PPARγ, activating the receptor to 25-30% of that produced by the full agonist rosiglitazone (EC50 = 4.5 ?M). Through these actions, telmisartan potently reduces blood pressure in various animal models of hypertension, diminishing cardiac hypertrophy, cardiovascular and renal risk, and glomerulosclerosis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only