D-Amygdalin, 98%, Thermo Scientific Chemicals
D-Amygdalin, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

D-Amygdalin, 98%, Thermo Scientific Chemicals

A benzylic glycoside that has been used as an antineoplastic agent
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
25 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number J61472.06
also known as J61472-06
Price (USD)/ Each
89.60
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
89.60
Add to cart
D-Amygdalin, 98%, Thermo Scientific Chemicals
Catalog numberJ61472.06
Price (USD)/ Each
89.60
-
Add to cart
Chemical Identifiers
CAS29883-15-6
IUPAC Name2-phenyl-2-{[(2R,3R,4S,5S,6R)-3,4,5-trihydroxy-6-({[(2R,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy}methyl)oxan-2-yl]oxy}acetonitrile
Molecular FormulaC20H27NO11
InChI KeyXUCIJNAGGSZNQT-ARGBPYSKNA-N
SMILESOC[C@H]1O[C@@H](OC[C@H]2O[C@@H](OC(C#N)C3=CC=CC=C3)[C@H](O)[C@@H](O)[C@@H]2O)[C@H](O)[C@@H](O)[C@@H]1O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)E. coli, Salmonella, S. Aureus: Negative
Assay (HPLC)≥ 98 %
Assay (unspecified)Bacterial Endotoxin: ≤ 1000 cfu.gm
Assay (unspecified)Yeast and Mold: ≤ 10 cfu/gm
Heavy Metals≤ 5 ppm
View more
A benzylic glycoside that has been used as an antineoplastic agent

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only