Thermo Scientific Chemicals
Protease Inhibitor Cocktail, for mammalian cells
Catalog number: J61473.XF
1 mL, Each
Thermo Scientific Chemicals
Protease Inhibitor Cocktail, for mammalian cells
Catalog number: J61473.XF
1 mL, Each
Quantity
Catalog number: J61473.XF
also known as J61473-XF
Price (USD)
Have Questions?
Concentration
AEBSF: 104 mM
Concentration
Aprotinin: 80 µM
Concentration
Bestatin: 5 mM
Concentration
E-64: 1.5 mM
Concentration
Leupeptin: 2 mM