Thermo Scientific Chemicals

SB 203580, 98%

Catalog number: J61482.MF
50 mg, Each
Thermo Scientific Chemicals

SB 203580, 98%

Catalog number: J61482.MF
50 mg, Each
Quantity
Catalog number: J61482.MF
also known as J61482-MF
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
152121-47-6
IUPAC Name
4-[4-(4-fluorophenyl)-2-(4-methanesulfinylphenyl)-1H-imidazol-5-yl]pyridine
Molecular Formula
C21H16FN3OS
InChI Key
CDMGBJANTYXAIV-UHFFFAOYNA-N
SMILES
CS(=O)C1=CC=C(C=C1)C1=NC(=C(N1)C1=CC=NC=C1)C1=CC=C(F)C=C1
Form
Crystalline powder or powder or granular
Assay from Suppliers CofA
≥97.5%
Appearance (Color)
White to pale cream to pale yellow

Description

Highly specific new inhibitor of p38 kinase and MAP Kinase homologues

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text