Informational:Informational:This product is animal origin and can only be shipped within the United States
Thermo Scientific Chemicals
Actin, from Rabbit Muscle, Thermo Scientific Chemicals
Catalog number: J61490.MCR
1mg, Each
Thermo Scientific Chemicals
Actin, from Rabbit Muscle, Thermo Scientific Chemicals
Catalog number: J61490.MCR
1mg, Each
Quantity
Catalog number: J61490.MCR
also known as J61490-M^
Price (USD)
Have Questions?
Assay (unspecified)
A280: Report value
Assay (unspecified)
SDS-PAGE: Complies
Description
Used in cell transport studies, and inhibits deoxyribonuclease I