Amphotericin B, Streptomyces nodosus
Amphotericin B, Streptomyces nodosus
Amphotericin B, Streptomyces nodosus
Amphotericin B, Streptomyces nodosus
Thermo Scientific Chemicals

Amphotericin B, Streptomyces nodosus

Amphotericin B, CAS # 1397-89-3, is an antifungal that has been used against fungus caused by protozoan parasites of Leishmania genus. | CAS: 1397-89-3 | C47H73NO17 | 924.09 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
100 mg
5 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number J61491.03
also known as J61491-03
Price (USD)
189.65
Online Exclusive
211.00
Save 21.35 (10%)
Each
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)
189.65
Online Exclusive
211.00
Save 21.35 (10%)
Each
Add to cart
Chemical Identifiers
CAS1397-89-3
IUPAC Name(1R,3S,5R,6R,9R,11R,15S,16R,17R,18S,19Z,21Z,23Z,25Z,27Z,29Z,31Z,33R,35S,36R,37S)-33-{[(2R,3S,4S,5S,6R)-4-amino-3,5-dihydroxy-6-methyloxan-2-yl]oxy}-1,3,5,6,9,11,17,37-octahydroxy-15,16,18-trimethyl-13-oxo-14,39-dioxabicyclo[33.3.1]nonatriaconta-19,21,23,25,27,29,31-heptaene-36-carboxylic acid
Molecular FormulaC47H73NO17
InChI KeyAPKFDSVGJQXUKY-ZNVUZQDLSA-N
SMILESC[C@H]1O[C@@H](O[C@@H]2C[C@@H]3O[C@@](O)(C[C@H](O)[C@H]3C(O)=O)C[C@@H](O)C[C@@H](O)[C@H](O)CC[C@@H](O)C[C@@H](O)CC(=O)O[C@@H](C)[C@H](C)[C@H](O)[C@@H](C)\C=C/C=C\C=C/C=C\C=C/C=C\C=C/2)[C@@H](O)[C@@H](N)[C@@H]1O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)Amphotericin A: 15.0% max.
Assay (unspecified)Potency: 750 ?g/mg min.
Loss on Drying5.0% max.
Ask our AI about this Product
Amphotericin B, antifungal activity has been used against leishmaniasis caused by protozoan parasites of the Leishmania genus. Amphotericin B trihydrate was observed to cause suppression of bone marrow progenitor cells and to induce Actinomycin D(CR001) sensitivity in drug resistant HELA cells in vitro.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

  • Amphotericin B is an antibiotic that has antifungal activity and is produced mainly by Streptomyces nodosus

Applications

  • It has antifungal activity against fungus caused by protozoan parasites of Leishmania genus by binding to an essential component of the fungal cell membrane and altering cell membrane permeability
  • It can suppress bone marrow progenitor cells and induce Actinomycin D(CR001) sensitivity in drug resistant HELA cells in vitro
RUO – Research Use Only

General References:

  1. Boulware, R.D. et. al. Timing of antiretroviral therapy after diagnosis of cryptococcal meningitis. New England Journal of Medicine. 2014, 370, (26), 2487-2498.
  2. Sarkar, S. et. al. Therapeutic activation of macrophages and microglia to suppress brain tumor-initiating cells. Nature Neuroscience. 2014, 17, (1), 46-55.
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link