BioAcryl-P (30%, 37.5:1), Thermo Scientific Chemicals
BioAcryl-P (30%, 37.5:1), Thermo Scientific Chemicals
Thermo Scientific Chemicals

BioAcryl-P (30%, 37.5:1), Thermo Scientific Chemicals

BioAcryl-P (Acrylamide/Bisacrylamide 37.5:1, CAS # 25034-58-6) is a powder-based formula used to create the gel matrix necessary for electrophoresis experiments.
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
500 mL
Catalog number J61505.K2
also known as J61505-K2
Price (USD)/ Each
232.65
Online exclusive
258.00 
Save 25.35 (10%)
-
Add to cart
Quantity:
1 L
Request bulk or custom format
Price (USD)/ Each
232.65
Online exclusive
258.00 
Save 25.35 (10%)
Add to cart
BioAcryl-P (30%, 37.5:1), Thermo Scientific Chemicals
Catalog numberJ61505.K2
Price (USD)/ Each
232.65
Online exclusive
258.00 
Save 25.35 (10%)
-
Add to cart
SpecificationsSpecification SheetSpecification Sheet
Chemical Name or MaterialAcrylamide/Bisacrylamide 37.5:1
Physical FormLiquid
Health Hazard 1H302+H332-H315-H317-H319-H340-H350-H361f-H372
Health Hazard 2GHS H Statement
H340-H350-H372-H361-H302-H315-H319-H317
May cause genetic defects.
May cause cancer.
Causes damage to organs through prolonged or repeated exposure.
Suspected of damaging fertility or the unborn child.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
Health Hazard 3P201-P202-P260-P264b-P270-P271-P272-P280-P281-P301+P312-P302+P352-P304+P340-P305+P351+P338-P308+P313-P330-P333+P313-P362-P501c
View more
Acrylamide/Bisacrylamide 37.5:1, Premixed powder is used for preparing protein DNA gels. It is used as a DNA sequencing gels, Protein separation. They are also used for preparing gels using stock solutions with the most common cross-link ratios.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

  • BioAcryl-P is a 37.5:1 ratio of Acrylamide/Bisacrylamide
  • Acrylamide is a crystalline amide that forms polymer structures at room temperature
  • Acrylamide acts as the monomer for the gel structure, and bisacrylamide as the crosslinker for gel matrix formation

Application

  • Suitable for gel electrophoresis-mediated separation of proteins and DNA for identification or sequencing
WARNING: Cancer and Reproductive Harm – www.P65Warnings.ca.gov
RUO – Research Use Only

General References:

  1. H Eshaghpour.; DM Crothers. Preparative separation of the complementary strands of DNA restriction fragments by alkaline RPC-5 chromatography. Nucl. Acids Res. 1978, 5,(1), 13-21.
  2. M Bazile, HA Nichols, JA Pojman. Effect of orientation on thermoset frontal polymerization. Journal of Polymer. 2002, 40,(20), 3504-3508.