Bradford Dye Reagent, Ready-to-Use soln.
Bradford Dye Reagent, Ready-to-Use soln.
Thermo Scientific Chemicals

Bradford Dye Reagent, Ready-to-Use soln.

Suitable for protein concentration determination (100 to 1500μg)
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
1 L
Catalog number J61522.AP
also known as J61522-AP
Price (USD)/ Each
91.65
Online exclusive
102.00 
Save 10.35 (10%)
-
Add to cart
Quantity:
500 mL
Request bulk or custom format
Price (USD)/ Each
91.65
Online exclusive
102.00 
Save 10.35 (10%)
Add to cart
Bradford Dye Reagent, Ready-to-Use soln.
Catalog numberJ61522.AP
Price (USD)/ Each
91.65
Online exclusive
102.00 
Save 10.35 (10%)
-
Add to cart
SpecificationsSpecification SheetSpecification Sheet
Chemical Name or MaterialBradford Dye Reagent
ColorBlue
Name NoteReady-to-Use soln.
Health Hazard 1H290-H302+H332-H314-H335-H371
Health Hazard 2GHS H Statement
H314-H318-H371
Causes severe skin burns and eye damage.
Causes serious eye damage.
May cause damage to organs.
View more
The Bradford Reagent is suitable for protein concentration determination (100-1500μg).Bradford dye reagent is used in the determination of protein content in cell fractions and assessing the protein concentrations.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only