Thermo Scientific Chemicals

Cyclopamine, 99%, Thermo Scientific Chemicals

Catalog number: J61528.MB
25 mg, Each
Thermo Scientific Chemicals

Cyclopamine, 99%, Thermo Scientific Chemicals

Catalog number: J61528.MB
25 mg, Each
Quantity
Catalog number: J61528.MB
also known as J61528-MB
Price (USD)
Quantity
-
Have Questions?

Description

Cyclopamine, is used as a hedgehog [Hh] signaling pathway and Smo inhibitor. It depletes stem-like cancer cells in glioblastoma and blocks tumor engraftment. Can also be used to induce differentiation of human embryonic stem cells (hESCs) into hormone-expressing endocrine cells.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cyclopamine, is used as a hedgehog [Hh] signaling pathway and Smo inhibitor. It depletes stem-like cancer cells in glioblastoma and blocks tumor engraftment. Can also be used to induce differentiation of human embryonic stem cells (hESCs) into hormone-expressing endocrine cells.

Solubility
Soluble in ethanol (5 mg/ml warm), DMSO (4 mg/ml), DMF (10 mg/ml), methylene chloride, chloroform, methanol (7 mg/ml), and benzene.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature: 2 - 8°C. Keep away from strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text