Thermo Scientific Chemicals

Tween 20 blocking buffer, 1% in PBS (10X)

Catalog number: J61544.AP
500 mL, Each
Thermo Scientific Chemicals

Tween 20 blocking buffer, 1% in PBS (10X)

Catalog number: J61544.AP
500 mL, Each
Quantity
Catalog number: J61544.AP
also known as J61544-AP
Price (USD)
Quantity
-
Have Questions?
Appearance (Color)
Clear colorless
Form
Liquid
Composition
101 mM Sodium phosphate dibasic, 17.6 mM Potassium phosphate monobasic, 27 mM Potassium chloride, 1.37 M Sodium chloride, 1% Tween 20
Comment
Prepared in 18.2megohms-cm ± 1 water, autoclaved for 30 min on liquid cycle at 15 psi and 121 degress C
pH
7.4 ± 0.20 (1X)

Description

Tween 20 blocking buffer solution is used as a washing agent in immunoassays, such as western blots and ELISAs. It helps to prevent non-specific antibody binding. The buffer is used for washes between each immunoreactions, to remove unbound immunological, and eventually for incubation solutions of immunoreagents to reduce unspecific background.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text