Tween 20 blocking buffer, 1% in PBS (10X)
Tween 20 blocking buffer, 1% in PBS (10X)
Thermo Scientific Chemicals

Tween 20 blocking buffer, 1% in PBS (10X)

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
500 mL
2 L
Catalog number J61544.K2
also known as J61544-K2
Price (USD)/ Each
102.00
-
Add to cart
Quantity:
1 L
Request bulk or custom format
Price (USD)/ Each
102.00
Add to cart
Tween 20 blocking buffer, 1% in PBS (10X)
Catalog numberJ61544.K2
Price (USD)/ Each
102.00
-
Add to cart
SpecificationsSpecification SheetSpecification Sheet
Product TypeBlocking Buffer
Quantity1 L
FormViscous Liquid
Recommended StorageAmbient temperatures
Concentration1% in PBS (10X)
View more
Tween 20 blocking buffer solution is used as a washing agent in immunoassays, such as western blots and ELISAs. It helps to prevent non-specific antibody binding. The buffer is used for washes between each immunoreactions, to remove unbound immunological, and eventually for incubation solutions of immunoreagents to reduce unspecific background.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only
Contents & Storage
2°C to 8°C