Thermo Scientific Chemicals

Metoclopramide hydrochloride, Thermo Scientific Chemicals

Catalog number: J61545.09
10 g, Each
Thermo Scientific Chemicals

Metoclopramide hydrochloride, Thermo Scientific Chemicals

Catalog number: J61545.09
10 g, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: J61545.09
also known as J61545-09
Price (USD)
Quantity
-
Have Questions?

Description

Metoclopramide hydrochloride is used as a digestive system drugs. It is used for anti-emetic, used for digestive dysfunction.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Metoclopramide hydrochloride is used as a digestive system drugs. It is used for anti-emetic, used for digestive dysfunction.

Solubility
It is very soluble in water, easily soluble in ethyl alcohol, slightly soluble in dichloromethane.

Notes
Hygroscopic, Refrigerator, Under Inert atmosphere. Incompatible to strong alkalis, strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text