Thermo Scientific Chemicals

Potassium phosphate, 0.5M buffer soln., pH 7.5

Catalog number: J61553.AP
500 mL, Each
Thermo Scientific Chemicals

Potassium phosphate, 0.5M buffer soln., pH 7.5

Catalog number: J61553.AP
500 mL, Each
Quantity
Catalog number: J61553.AP
also known as J61553-AP
Price (USD)
Quantity
-
Have Questions?

Description

Potassium phosphate buffer is typically used as a component for a wide variety of media used in the culture of microorganisms, as a component in phosphate buffered saline. In addition to helping maintain pH, it supplies essential phosphate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Potassium phosphate buffer is typically used as a component for a wide variety of media used in the culture of microorganisms, as a component in phosphate buffered saline. In addition to helping maintain pH, it supplies essential phosphate.

Solubility
Soluble in water, Insoluble in ethanol, alcohol.

Notes
Store sealed bottle containing the dehydrated medium at 2 - 30C. Once opened and recapped, place container in a low humidity environment at the same storage temperature. Protect from moisture and light by keeping container tightly closed.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text