Thermo Scientific Chemicals

Mianserin hydrochloride, Thermo Scientific Chemicals

Catalog number: J61570.MC
100 mg, Each
Thermo Scientific Chemicals

Mianserin hydrochloride, Thermo Scientific Chemicals

Catalog number: J61570.MC
100 mg, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: J61570.MC
also known as J61570-MC
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
21535-47-7
IUPAC Name
hydrogen 5-methyl-2,5-diazatetracyclo[13.4.0.0²,⁷.0⁸,¹³]nonadeca-1(19),8,10,12,15,17-hexaene chloride
Molecular Formula
C18H21ClN2
InChI Key
YNPFMWCWRVTGKJ-UHFFFAOYNA-N
SMILES
[H+].[Cl-].CN1CCN2C(C1)C1=CC=CC=C1CC1=CC=CC=C21
Purity
≥98.0%
Form
Powder

Description

Mianserin hydrochloride is used as an antidepressant, antagonist, inverse agonist at 5-HT2 serotonin receptors, also blocks the H1 histamine receptor and the α2 adrenoceptor.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Mianserin hydrochloride is used as an antidepressant, antagonist, inverse agonist at 5-HT2 serotonin receptors, also blocks the H1 histamine receptor and the α2 adrenoceptor.

Solubility
Soluble in water, ethano
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text