RNase Removal Reagent
RNase Removal Reagent
Thermo Scientific Chemicals

RNase Removal Reagent

 
Have Questions?
Quantity:
475 mL
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number J61574.REX
also known as J61574-R!
Price (USD)/ Each
-
Quantity:
475 mL
Request bulk or custom format
Specifications
ColorColorless
Recommended StorageAmbient temperatures
Density1.00
For Use With (Application)Destroys RNases and DNases on contact
Product TypeRNase Removal Reagent
View more
Destroys RNases and DNases on contactRNase removal reagent is used to remove the RNase from glasswares, plastic surfaces, equipments, gel boxes, benchtops and labware. It is also used to eliminate the RNase from microcentrifuge tubes without any side enzymatic reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only