Thermo Scientific Chemicals

Calciferol, Thermo Scientific Chemicals

Catalog number: J61610.03
1 g, Each
Thermo Scientific Chemicals

Calciferol, Thermo Scientific Chemicals

Catalog number: J61610.03
1 g, Each
Quantity
Catalog number: J61610.03
also known as J61610-03
Price (USD)
Have Questions?

Chemical Identifiers

CAS
50-14-6
Appearance (Color)
White
Melting Point
114.0-120.0°C
Assay (unspecified)
≥97.0%
Form
Powder or crystals or crystalline powder
Optical Rotation
+102 to + 107°

Description

Calciferol is a strong inhibitory effect against bladder tumor promotion by sodium saccharin and induces cell differentiation in leukemia cells. Calciferol also been shown to be an inhibitor of DNA polymerase.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Calciferol is a strong inhibitory effect against bladder tumor promotion by sodium saccharin and induces cell differentiation in leukemia cells. Calciferol also been shown to be an inhibitor of DNA polymerase.

Solubility
Soluble in ethanol, water, methanol, dimethylformamide, and dimethyl sulfoxide.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text