Erbstatin Analog
Thermo Scientific Chemicals

Erbstatin Analog

An inhibitor of the EGF receptor associated tyrosine kinase
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 mg
10 mg
Catalog number J61612.LB0
also known as J61612-#0
Price (USD)/ Each
87.50
-
Add to cart
Quantity:
5 mg
Request bulk or custom format
Price (USD)/ Each
87.50
Add to cart
Erbstatin Analog
Catalog numberJ61612.LB0
Price (USD)/ Each
87.50
-
Add to cart
Specifications
Quantity5 mg
DescriptionYellow
For Use With (Application)An inhibitor of the EGF receptor associated tyrosine kinase
Recommended StorageStore at -20°C
CAS63177-57-1
View more
An inhibitor of the EGF receptor associated tyrosine kinase

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only
Contents & Storage
-30°C to -10°C