5,8,11,14-Eicosatetraynoic acid, Thermo Scientific Chemicals
5,8,11,14-Eicosatetraynoic acid, Thermo Scientific Chemicals
Thermo Scientific Chemicals

5,8,11,14-Eicosatetraynoic acid, Thermo Scientific Chemicals

A cyclooxygenase, lipoxygenase and cytochrome P450 inhibitor
Have Questions?
Change viewbuttonViewtableView
Quantity:
20 mg
100 mg
Catalog number J61624.FPL
also known as J61624-F+
Price (USD)/ Each
176.00
-
Add to cart
Quantity:
20 mg
Request bulk or custom format
Price (USD)/ Each
176.00
Add to cart
5,8,11,14-Eicosatetraynoic acid, Thermo Scientific Chemicals
Catalog numberJ61624.FPL
Price (USD)/ Each
176.00
-
Add to cart
Chemical Identifiers
CAS1191-85-1
IUPAC Nameicosa-5,8,11,14-tetraynoic acid
Molecular FormulaC20H24O2
InChI KeyMGLDCXPLYOWQRP-UHFFFAOYSA-N
SMILESCCCCCC#CCC#CCC#CCC#CCCCC(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Off-white
FormSolid
Purity≥97.0%
A cyclooxygenase, lipoxygenase and cytochrome P450 inhibitor

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only