Thermo Scientific Chemicals

Transferrin (Apo), bovine plasma, 98+%

Catalog number: J61626.MC
100mg, Each
Thermo Scientific Chemicals

Transferrin (Apo), bovine plasma, 98+%

Catalog number: J61626.MC
100mg, Each
Quantity
Catalog number: J61626.MC
also known as J61626-MC
Price (USD)
Have Questions?
Assay (unspecified)
(Beta Globulins 3% solution) Cellulose Acetate Electrophoresis: 95-100%
Assay (unspecified)
Endotoxin by Limulus Amebocyte Lysate: 0-1.0 EU/mg
Water Content (Karl Fischer Titration)
< 5.0%
pH
7.0-8.0

Description

Apo-Transferrin is also an important component used for the cultivation of mammalian cells in-vitro because it acts as a transport factor for defined culture media. This property is crucial for long-term mammalian cell growth in-vitro since ATF operates in media by binding to contaminating metal ions. Some other common uses for Apo-Transferrin are affinity purification and receptor mediated transfection of DNA-like molecules into cells.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text