(-)-Epigallocatechin, Thermo Scientific Chemicals
(-)-Epigallocatechin, Thermo Scientific Chemicals
Thermo Scientific Chemicals

(-)-Epigallocatechin, Thermo Scientific Chemicals

 
Have Questions?
Quantity:
10 mg
Catalog number J61630.MA
also known as J61630-MA
Price (USD)/ Each
170.65
Online exclusive
189.00 
Save 18.35 (10%)
-
Add to cart
Quantity:
10 mg
Request bulk or custom format
Price (USD)/ Each
170.65
Online exclusive
189.00 
Save 18.35 (10%)
Add to cart
(-)-Epigallocatechin, Thermo Scientific Chemicals
Catalog numberJ61630.MA
Price (USD)/ Each
170.65
Online exclusive
189.00 
Save 18.35 (10%)
-
Add to cart
Chemical Identifiers
CAS970-74-1
IUPAC Name(2R,3R)-2-(3,4,5-trihydroxyphenyl)-3,4-dihydro-2H-1-benzopyran-3,5,7-triol
Molecular FormulaC15H14O7
InChI KeyXMOCLSLCDHWDHP-IUODEOHRSA-N
SMILESO[C@@H]1CC2=C(O)C=C(O)C=C2O[C@@H]1C1=CC(O)=C(O)C(O)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Purity≥98.0%
FormPowder
A natural product from green tea that induces apoptosis

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only