Adenosine-5'-monophosphate disodium salt, Thermo Scientific Chemicals
Adenosine-5'-monophosphate disodium salt, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Adenosine-5'-monophosphate disodium salt, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number J61643.06
also known as J61643-06
Price (USD)/ Each
31.65
Online exclusive
34.80 
Save 3.15 (9%)
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
31.65
Online exclusive
34.80 
Save 3.15 (9%)
Add to cart
Adenosine-5'-monophosphate disodium salt, Thermo Scientific Chemicals
Catalog numberJ61643.06
Price (USD)/ Each
31.65
Online exclusive
34.80 
Save 3.15 (9%)
-
Add to cart
Chemical Identifiers
CAS4578-31-8
IUPAC Namedisodium [(2R,3S,4R,5R)-5-(6-amino-9H-purin-9-yl)-3,4-dihydroxyoxolan-2-yl]methyl phosphate
Molecular FormulaC10H12N5Na2O7P
InChI KeyQGXLVXZRPRRCRP-IDIVVRGQSA-L
SMILES[Na+].[Na+].NC1=C2N=CN([C@@H]3O[C@H](COP([O-])([O-])=O)[C@@H](O)[C@H]3O)C2=NC=N1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale cream to pale yellow
FormPowder or crystals or crystalline powder
Assay from Supplier's CofA≥98.0% (HPLC)
Water Content (Karl Fischer Titration)≤26.0%
Adenosine-5'-monophosphate disodium salt is a nucleotide involved in the reactions of cellular energy transfers. It is useful for skin rejuvenation in dermatology and aesthetic dermatology. It acts as an activator of a class of protein kinases called as AMP-activated protein kinase (AMPK). Further, it is used as a substrate for enzymes such as AMP-thymidine kinase, AMP deaminase and 5'-nucleotidase.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Adenosine-5′-monophosphate disodium salt is a nucleotide involved in the reactions of cellular energy transfers. It is useful for skin rejuvenation in dermatology and aesthetic dermatology. It acts as an activator of a class of protein kinases called as AMP-activated protein kinase (AMPK). Further, it is used as a substrate for enzymes such as AMP-thymidine kinase, AMP deaminase and 5′-nucleotidase.

Solubility
Soluble in water, dimethyl sulfoxide and methanol.

Notes
Air and moisture sensitive. Hygroscopic. Incompatible with strong oxidizing agents.
RUO – Research Use Only

General References:

  1. Cade-Menun, B. J. Improved peak identification in 31P-NMR spectra of environmental samples with a standardized method and peak library. Geoderma 2015, 257-258, 102-114.
  2. Ortiz-Villanueva, E.; Jaumot, J.; Benavente, F.; Piña, B.; Sanz-Nebot, V.; Tauler, R. Combination of CE-MS and advanced chemometric methods for high-throughput metabolic profiling. Electrophoresis 2015, 36 (18), 2324-2335.