Thermo Scientific Chemicals

L-Cystine, Cell Culture Reagent, Thermo Scientific Chemicals

Catalog number: J61651.22
100 g, Each
Thermo Scientific Chemicals

L-Cystine, Cell Culture Reagent, Thermo Scientific Chemicals

Catalog number: J61651.22
100 g, Each
Quantity
Catalog number: J61651.22
also known as J61651-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
56-89-3
IUPAC Name
(2R)-2-amino-3-{[(2R)-2-amino-2-carboxyethyl]disulfanyl}propanoic acid
Molecular Formula
C6H12N2O4S2
InChI Key
LEVWYRKDKASIDU-IMJSIDKUSA-N
SMILES
N[C@@H](CSSC[C@H](N)C(O)=O)C(O)=O
Assay (unspecified)
≥95.0%
Infrared spectrum
Conforms
Appearance (Color)
White
Comment
Microbial Count: ≤1000 cfu/g (total plate count)
Loss on Drying
≤0.2%

Description

L-cysteine acts as a major source of sulfur in cell culture. Cysteine has been used in pulse-chase experiments to study protein expression from an inducible promoter. L-Cysteine is used to supplement cell culture media.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
L-cysteine acts as a major source of sulfur in cell culture. Cysteine has been used in pulse-chase experiments to study protein expression from an inducible promoter. L-Cysteine is used to supplement cell culture media.

Solubility
It is soluble in water and HCL.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from heat.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text