Thermo Scientific Chemicals

13-cis-Retinoic acid, Thermo Scientific Chemicals

Catalog number: J61666.ME
500 mg, Each
Thermo Scientific Chemicals

13-cis-Retinoic acid, Thermo Scientific Chemicals

Catalog number: J61666.ME
500 mg, Each
Quantity
Catalog number: J61666.ME
also known as J61666-ME
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
4759-48-2
IUPAC Name
(2Z,4E,6E,8E)-3,7-dimethyl-9-(2,6,6-trimethylcyclohex-1-en-1-yl)nona-2,4,6,8-tetraenoic acid
Molecular Formula
C20H28O2
InChI Key
SHGAZHPCJJPHSC-XFYACQKRSA-N
SMILES
CC(C=CC1=C(C)CCCC1(C)C)=C/C=C/C(/C)=CC(O)=O
Assay (HPLC)
98.0% min.
Appearance (Color)
Yellow to orange
Form
Solid

Description

13-cis-Retinoic acid is used to inhibits cell proliferation and induces cell differentiation. 13-cis-Retinoic acid is primarily used in the treatment of severe recalcitrant nodular acne.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in chloroform, dimethyl sulfoxide, water, ethanol, and ether.

Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text