Thermo Scientific Chemicals

Monensin sodium salt, 90% min., Thermo Scientific Chemicals

Catalog number: J61669.03
1 g, Each
Thermo Scientific Chemicals

Monensin sodium salt, 90% min., Thermo Scientific Chemicals

Catalog number: J61669.03
1 g, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: J61669.03
also known as J61669-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
22373-78-0
IUPAC Name
sodium (2S,3R,4S)-4-[(2S,5R,7S,8R,9S)-2-[(2S,2'R,3'S,5R,5'R)-2-ethyl-5'-[(2S,3S,5R,6R)-6-hydroxy-6-(hydroxymethyl)-3,5-dimethyloxan-2-yl]-3'-methyl-[2,2'-bioxolan]-5-yl]-9-hydroxy-2,8-dimethyl-1,6-dioxaspiro[4.5]decan-7-yl]-3-methoxy-2-methylpentanoate
Molecular Formula
C36H61NaO11
InChI Key
XOIQMTLWECTKJL-FBZUZRIGSA-M
SMILES
[Na+].CC[C@]1(CC[C@@H](O1)[C@]1(C)CC[C@]2(C[C@H](O)[C@@H](C)[C@H](O2)[C@@H](C)[C@@H](OC)[C@H](C)C([O-])=O)O1)[C@@H]1O[C@H](C[C@@H]1C)[C@H]1O[C@@](O)(CO)[C@H](C)C[C@@H]1C
Form
Powder
Assay (unspecified)
Purity: 90.0% min.
Appearance (Color)
White to off-white

Description

Monensin sodium salt is used in potentiometric and spectroscopic studies of alkali metal ion complexes. Monensin (Na) is a polyether small molecule ionophore, capable of forming stable complexes to monovalent cations with specific affinity for Na+. Monensin presents a highly lipophilic scaffold around the bound ion, allowing movement of the complex through the lipid bilayer and consequent transport of the ion out of the cell. Excessive flux of ions out of the cell produces a cytotoxic effect and generates the antibiotic properties of Monensin.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Monensin sodium salt is used in potentiometric and spectroscopic studies of alkali metal ion complexes. Monensin (Na) is a polyether small molecule ionophore, capable of forming stable complexes to monovalent cations with specific affinity for Na+. Monensin presents a highly lipophilic scaffold around the bound ion, allowing movement of the complex through the lipid bilayer and consequent transport of the ion out of the cell. Excessive flux of ions out of the cell produces a cytotoxic effect and generates the antibiotic properties of Monensin.

Solubility
Soluble in methanol at 50mg/ml

Notes
Research Use only: Not intended for animal or human diagnostic or therapeutic use.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text