Haloperidol, Thermo Scientific™
Haloperidol, Thermo Scientific™
Thermo Scientific™

Haloperidol, Thermo Scientific™

Have Questions?
Quantity:
5 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number J61688.06
also known as J61688-06
Price (USD)/ Each
109.00
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
109.00
Add to cart
Haloperidol, Thermo Scientific™
Catalog numberJ61688.06
Price (USD)/ Each
109.00
-
Add to cart
Chemical Identifiers
CAS52-86-8
IUPAC Name4-[4-(4-chlorophenyl)-4-hydroxypiperidin-1-yl]-1-(4-fluorophenyl)butan-1-one
Molecular FormulaC21H23ClFNO2
InChI KeyLNEPOXFFQSENCJ-UHFFFAOYSA-N
SMILESOC1(CCN(CCCC(=O)C2=CC=C(F)C=C2)CC1)C1=CC=C(Cl)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Melting Point149.0-154.0°C
FormPowder or crystals or crystalline powder
Appearance (Color)White to off-white
Assay from Supplier's CofA≥97.0% (HPLC)
An antagonist for D2, D3, and D4 dopamine receptors that induces apoptosis of neurons in the striatum of rats

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

An antagonist for D2, D3, and D4 dopamine receptors that induces apoptosis of neurons in the striatum of rats.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only