Thermo Scientific Chemicals

Harmaline, 98+%, Thermo Scientific Chemicals

Catalog number: J61699.06
5 g, Each
Thermo Scientific Chemicals

Harmaline, 98+%, Thermo Scientific Chemicals

Catalog number: J61699.06
5 g, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: J61699.06
also known as J61699-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
304-21-2
IUPAC Name
7-methoxy-1-methyl-2H,3H,4H-pyrido[3,4-b]indole
Molecular Formula
C13H14N2O
InChI Key
QJOZJXNKVMFAET-UHFFFAOYSA-N
SMILES
COC1=CC2=NC3=C(C)NCCC3=C2C=C1
Melting Point
237 - 243?C
Form
Crystalline Powder
Solution Test
Soluble in DMSO
Assay (HPLC)
≥ 98%
Water Content (Karl Fischer Titration)
≤ 1.0%

Description

CNS stimulant; may act through NMDA receptors. Reversible inhibitor of MAO-A

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • CNS stimulant; may act through NMDA receptors
  • Reversible inhibitor of MAO-A
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text