Thermo Scientific Chemicals

Prazosin hydrochloride, Thermo Scientific Chemicals

Catalog number: J61712.MD
250 mg, Each
Thermo Scientific Chemicals

Prazosin hydrochloride, Thermo Scientific Chemicals

Catalog number: J61712.MD
250 mg, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: J61712.MD
also known as J61712-MD
Price (USD)
Quantity
-
Have Questions?

Description

Prazosin hydrochloride is an inhibitor of PDE. Also offered in a free base form Prazosin

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Prazosin hydrochloride is an inhibitor of PDE. Also offered in a free base form Prazosin

Solubility
Soluble in dimethyl sulfoxide, ethanol, water, methanol.

Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text