General Description
• Tetracycline is a semisynthetic compound derived from chlortetracycline, an isolate from Streptomyces aureofaciens
• Tetracycline is a broad-spectrum bacteriostatic that inhibits polypeptide formation by binding the 30S ribosomal subunit and preventing aminoacyl tRNA-mRNA complex formation
• It may have other bactericidal properties such as limited binding to the 50S subunit
Application
• Tetracycline has been shown to exhibit antiprotozoal properties
• It can be used to kill both gram-positive and gram-negative bacteria
• It is suitable for use in laboratory applications in resistance gene testing and research on infection
Literature References
Armstrong, A.W.; Hekmatjah, J.; Kircik, L.H. Oral Tetracyclines and Acne: A Systematic Review for Dermatologists. J Drugs Dermatol. 2020, 19(11), s6-s13.
Li, W.Q.; Drucker, A.M.; Cho, E.; Laden, F.; VoPham, T., Li, S., Weinstock, M.A.; Qureshi, A.A. Tetracycline use and risk of incident skin cancer: a prospective study. Br J Cancer. 2018, 118(2), 294-298.