Ethylene glycol-O,O'-bis(2-aminoethyl)-N,N,N',N'-tetraacetic acid, 0.5M aq. soln., pH 8.0, autoclaved, Thermo Scientific Chemicals
Ethylene glycol-O,O'-bis(2-aminoethyl)-N,N,N',N'-tetraacetic acid, 0.5M aq. soln., pH 8.0, autoclaved, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Ethylene glycol-O,O'-bis(2-aminoethyl)-N,N,N',N'-tetraacetic acid, 0.5M aq. soln., pH 8.0, autoclaved, Thermo Scientific Chemicals

 
Have Questions?
Quantity:
50 mL
Catalog number J61721.AD
also known as J61721-AD
Price (USD)/ Each
96.65
Online exclusive
107.00 
Save 10.35 (10%)
-
Add to cart
Quantity:
50 mL
Request bulk or custom format
Price (USD)/ Each
96.65
Online exclusive
107.00 
Save 10.35 (10%)
Add to cart
Ethylene glycol-O,O'-bis(2-aminoethyl)-N,N,N',N'-tetraacetic acid, 0.5M aq. soln., pH 8.0, autoclaved, Thermo Scientific Chemicals
Catalog numberJ61721.AD
Price (USD)/ Each
96.65
Online exclusive
107.00 
Save 10.35 (10%)
-
Add to cart
Chemical Identifiers
CAS67-42-5
IUPAC Name3,12-bis(carboxymethyl)-6,9-dioxa-3,12-diazatetradecanedioic acid
Molecular FormulaC14H24N2O10
InChI KeyDEFVIWRASFVYLL-UHFFFAOYSA-N
SMILESOC(=O)CN(CCOCCOCCN(CC(O)=O)CC(O)=O)CC(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
pH8.00+/-0.20
FormLiquid
CommentFiltered through 0.22 - micron filter.
Concentration500 mM EGTA
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only