Thermo Scientific Chemicals

Dextromethorphan hydrobromide, Thermo Scientific Chemicals

Catalog number: J61732.18
50 g, Each
Thermo Scientific Chemicals

Dextromethorphan hydrobromide, Thermo Scientific Chemicals

Catalog number: J61732.18
50 g, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: J61732.18
also known as J61732-18
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
125-69-9
IUPAC Name
hydrogen (1S,9S,10S)-4-methoxy-17-methyl-17-azatetracyclo[7.5.3.0¹,¹⁰.0²,⁷]heptadeca-2,4,6-triene bromide
Molecular Formula
C18H26BrNO
InChI Key
MISZALMBODQYFT-URVXVIKDSA-N
SMILES
[H+].[Br-].COC1=CC=C2C[C@H]3[C@H]4CCCC[C@@]4(CCN3C)C2=C1
Appearance (Color)
White
Optical Rotation
+27? to +30? (c = 1.5 in water)
pH
5.2-6.5
Form
Crystalline powder
Assay from Supplier's CofA
≥98% (HPLC)

Description

It is used as a potentially useful NMDA receptor agonist. it is usually used as central nervous antitussive, cough suppressant. This is one of the most popular ingredients in over-the-counter cold and cough medications. Dextromethorphan hydrobromide may be added to other preparations to help treat pain and a variety of other disturbances as well. It is used as a pharmaceutical raw material.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as a potentially useful NMDA receptor agonist. it is usually used as central nervous antitussive, cough suppressant. This is one of the most popular ingredients in over-the-counter cold and cough medications. Dextromethorphan hydrobromide may be added to other preparations to help treat pain and a variety of other disturbances as well. It is used as a pharmaceutical raw material.

Solubility
Soluble in water (40 mM), and alcohol (1:10).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text