Thermo Scientific Chemicals

(-)-Epigallocatechin gallate, Thermo Scientific Chemicals

Catalog number: J61745.MF
50 mg, Each
Thermo Scientific Chemicals

(-)-Epigallocatechin gallate, Thermo Scientific Chemicals

Catalog number: J61745.MF
50 mg, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: J61745.MF
also known as J61745-MF
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
989-51-5
IUPAC Name
(2R,3R)-5,7-dihydroxy-2-(3,4,5-trihydroxyphenyl)-3,4-dihydro-2H-1-benzopyran-3-yl 3,4,5-trihydroxybenzoate
Molecular Formula
C22H18O11
InChI Key
WMBWREPUVVBILR-WIYYLYMNSA-N
SMILES
OC1=CC(O)=C2C[C@@H](OC(=O)C3=CC(O)=C(O)C(O)=C3)[C@H](OC2=C1)C1=CC(O)=C(O)C(O)=C1
UV/Vis
(|l max): 276nm
Thin Layer Chromatography
(Purity): 100%

Description

Epigallocatechin gallate is reported to strongly inhibit Cu(2+)-mediated oxidative modification of LDLs. EGCG also inhibits NOS2 (iNOS).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Epigallocatechin gallate is reported to strongly inhibit Cu(2+)-mediated oxidative modification of LDLs. EGCG also inhibits NOS2 (iNOS).

Solubility
Soluble in ethanol, dimethyl formamide, water.

Notes
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text