Thermo Scientific Chemicals

Calpain Inhibitor I, 95+%

Catalog number: J61766.LB0
5 mg, Each
Thermo Scientific Chemicals

Calpain Inhibitor I, 95+%

Catalog number: J61766.LB0
5 mg, Each
Quantity
Catalog number: J61766.LB0
also known as J61766-#0
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
110044-82-1
IUPAC Name
(2S)-2-[(2S)-2-acetamido-4-methylpentanamido]-4-methyl-N-[(2S)-1-oxohexan-2-yl]pentanamide
Molecular Formula
C20H37N3O4
InChI Key
FMYKJLXRRQTBOR-BZSNNMDCSA-N
SMILES
CCCC[C@H](NC(=O)[C@H](CC(C)C)NC(=O)[C@H](CC(C)C)NC(C)=O)C=O
Assay (HPLC)
95.0% min.

Description

It inhibits apoptosis in thymocytes and metamyelocytes. It also inhibits cell cycle progression at the G1/S border and metaphase/anaphase in CHO cells by inhibiting cyclin B degradation. ALLN inhibits the proteolysis of IκBα and IκBβ by the ubiquitin-proteasome complex. ALLN also prevents nitric oxide production by activated macrophages by interfering with transcription of the inducible nitric oxide synthase gene.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text