Thermo Scientific Chemicals

Erioglaucine sodium salt, Thermo Scientific Chemicals

Catalog number: J61780.06
5 g, Each
Thermo Scientific Chemicals

Erioglaucine sodium salt, Thermo Scientific Chemicals

Catalog number: J61780.06
5 g, Each
Quantity
Catalog number: J61780.06
also known as J61780-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
3844-45-9
IUPAC Name
disodium bis(4-{ethyl[(3-sulfonatophenyl)methyl]amino}phenyl)(2-sulfonatophenyl)methylium
Molecular Formula
C37H34N2Na2O9S3
InChI Key
SGHZXLIDFTYFHQ-UHFFFAOYSA-L
SMILES
[Na+].[Na+].CCN(CC1=CC=CC(=C1)S([O-])(=O)=O)C1=CC=C(C=C1)[C+](C1=CC=C(C=C1)N(CC)CC1=CC=CC(=C1)S([O-])(=O)=O)C1=CC=CC=C1S([O-])(=O)=O
Assay (unspecified)
Dye content: 65.0 % min.
Assay (unspecified)
Water insoluble matter: 0.20% max.
Trace Metals
40ppm max.

Description

Erioglaucine sodium salt is used as a colorant for foods and other substances to induce a color change.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Erioglaucine sodium salt is used as a colorant for foods and other substances to induce a color change.

Solubility
Soluble in water (30 mg/ml), ethanol.

Notes
Dye content: approx 60%
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text