Thermo Scientific Chemicals

Phenolphthalein, 2% soln. in 95% ethanol

Catalog number: J61785.AU
1000 mL, Each
Thermo Scientific Chemicals

Phenolphthalein, 2% soln. in 95% ethanol

Catalog number: J61785.AU
1000 mL, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: J61785.AU
also known as J61785-AU
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
77-09-8
IUPAC Name
3,3-bis(4-hydroxyphenyl)-1,3-dihydro-2-benzofuran-1-one
Molecular Formula
C20H14O4
InChI Key
KJFMBFZCATUALV-UHFFFAOYSA-N
SMILES
OC1=CC=C(C=C1)C1(OC(=O)C2=CC=CC=C12)C1=CC=C(O)C=C1
Form
Liquid
Appearance (Color)
Clear colorless
Assay (special)
Visual transition interval: Passes test

Description

It is used as a laboratory reagent and pH indicator. Phenolphthalein is a common indicator to use in neutralization titrations. Phenolphthalein exerts laxative effects by stimulating the intestinal mucosa and constricting smooth muscles. It is used medicinally as a cathartic.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Cancer and Reproductive Harm – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text