PIPES, 0.5M buffer soln., pH 6.8
PIPES, 0.5M buffer soln., pH 6.8
Thermo Scientific Chemicals

PIPES, 0.5M buffer soln., pH 6.8

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
100 mL
Catalog number J61786.AK
also known as J61786-AK
Price (USD)/ Each
132.65
Online exclusive
147.00 
Save 14.35 (10%)
-
Add to cart
Quantity:
250 mL
Request bulk or custom format
Price (USD)/ Each
132.65
Online exclusive
147.00 
Save 14.35 (10%)
Add to cart
PIPES, 0.5M buffer soln., pH 6.8
Catalog numberJ61786.AK
Price (USD)/ Each
132.65
Online exclusive
147.00 
Save 14.35 (10%)
-
Add to cart
Specifications
Chemical Name or MaterialPIPES buffer soln.
CAS5625-37-6
ColorColorless
Recommended StorageKeep cold
Concentration0.5 M
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only