Thermo Scientific Chemicals

Okadaic acid potassium salt, 98%, Thermo Scientific Chemicals

Catalog number: J61791.EXG
100 μg, Each
Thermo Scientific Chemicals

Okadaic acid potassium salt, 98%, Thermo Scientific Chemicals

Catalog number: J61791.EXG
100 μg, Each
Quantity
Catalog number: J61791.EXG
also known as J61791-!G
Price (USD)
Have Questions?

Description

Okadaic Acid, Potassium Salt has been used as a response modifier in the study of large granular lymphocyte and T-cell biochemistry. In addition to inhibitive activities it is a tumor promoter and has been used to study nitric oxide metabolism, apoptosis, cell cycle and calcium signaling. It is also used as a PP1/PP2A/PP2B/PP2C inhibitor, used to study tumor promotion/NO metabolism/apoptosis

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Okadaic Acid, Potassium Salt has been used as a response modifier in the study of large granular lymphocyte and T-cell biochemistry. In addition to inhibitive activities it is a tumor promoter and has been used to study nitric oxide metabolism, apoptosis, cell cycle and calcium signaling. It is also used as a PP1/PP2A/PP2B/PP2C inhibitor, used to study tumor promotion/NO metabolism/apoptosis

Solubility
Soluble in DMSO or ethanol. Soluble in water at 0.1mg/ml. Also soluble in methanol

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature: -20°C. Keep away from strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text