Thermo Scientific Chemicals

Cantharidin, 98%, Thermo Scientific Chemicals

Catalog number: J61801.MC
100mg, Each
Thermo Scientific Chemicals

Cantharidin, 98%, Thermo Scientific Chemicals

Catalog number: J61801.MC
100mg, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: J61801.MC
also known as J61801-MC
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
56-25-7
IUPAC Name
(1R,2S,6R,7S)-2,6-dimethyl-4,10-dioxatricyclo[5.2.1.0²,⁶]decane-3,5-dione
Molecular Formula
C10H12O4
InChI Key
DHZBEENLJMYSHQ-XCVPVQRUSA-N
SMILES
C[C@]12[C@@H]3CC[C@@H](O3)[C@@]1(C)C(=O)OC2=O
Form
Powder
Purity
≥97.0%

Description

Cantharidin is used as a potent inhibitor of PP2A phosphatase activity. It has been shown to stimulate cell cycle progression and induce premature mitosis, used topically (0.7%) as an anti-wart treatment, and has been shown to be active in cervical, tongue, neuroblastoma, bone, leukemia, ovarian, colon, and various other cancer cell lines.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cantharidin is used as a potent inhibitor of PP2A phosphatase activity. It has been shown to stimulate cell cycle progression and induce premature mitosis, used topically (0.7%) as an anti-wart treatment, and has been shown to be ac
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text