Thermo Scientific Chemicals

Minoxidil, Thermo Scientific Chemicals

Catalog number: J61803.06
5 g, Each
Thermo Scientific Chemicals

Minoxidil, Thermo Scientific Chemicals

Catalog number: J61803.06
5 g, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: J61803.06
also known as J61803-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
38304-91-5
IUPAC Name
6-amino-2-imino-4-(piperidin-1-yl)-1,2-dihydropyrimidin-1-ol
Molecular Formula
C9H15N5O
InChI Key
ZIMGGGWCDYVHOY-UHFFFAOYSA-N
SMILES
NC1=CC(=NC(=N)N1O)N1CCCCC1
Loss on Drying
0.5% max.
Assay (unspecified)
98.0% min.
Trace Metals
20ppm max.
Residue on ignition
0.5% max.

Description

Minoxidil is used as antihypertensive, antialopecia agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description
• Minoxidil is a piperidine that acts as a vasodilator when converted into its active metabolite, minoxidil sulphate
• Minoxidil opens adenosine triphosphate (ATP)-sensitive potassium channels in the plasma membrane and relaxes smooth muscle in arteriole walls. Relaxation of smooth muscle within the arterioles leads to decreased resistance and lower blood pressure
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text