Thermo Scientific Chemicals

MOPS, 1.0M buffer soln., pH 7.0

Catalog number: J61821.AP
500 mL, Each
Thermo Scientific Chemicals

MOPS, 1.0M buffer soln., pH 7.0

Catalog number: J61821.AP
500 mL, Each
Quantity
Catalog number: J61821.AP
also known as J61821-AP
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1132-61-2
IUPAC Name
4-(3-sulfonatopropyl)morpholin-4-ium
Molecular Formula
C7H15NO4S
InChI Key
DVLFYONBTKHTER-UHFFFAOYSA-N
SMILES
[O-]S(=O)(=O)CCC[NH+]1CCOCC1
Form
Liquid
pH
7.0 ± 0.15
Comment
Dissolved in UV treated 18.2 megohm-cm ± 1 water, followed by pH adjustment and filtered through 0.22 micron filter

Description

MOPS is frequently used as a buffering agent in biology and biochemistry. It used widely for polyacrylamide gel electrophoresis. It is used as a A morpholino propanesulfonic acid buffer.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
MOPS is frequently used as a buffering agent in biology and biochemistry. It used widely for polyacrylamide gel electrophoresis. It is used as a A morpholino propanesulfonic acid buffer.

Solubility
Soluble in water.

Notes
Usage above 20 mM in mammalian cell culture work is not recommended. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from Strong oxidizing agents, Strong bases.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text