Tolnaftate, Thermo Scientific Chemicals
Tolnaftate, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Tolnaftate, Thermo Scientific Chemicals

Antifungal agent
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number J61834.03
also known as J61834-03
Price (USD)/ Each
53.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
63.00 
Save 9.35 (15%)
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
53.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
63.00 
Save 9.35 (15%)
Add to cart
Tolnaftate, Thermo Scientific Chemicals
Catalog numberJ61834.03
Price (USD)/ Each
53.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
63.00 
Save 9.35 (15%)
-
Add to cart
Chemical Identifiers
CAS2398-96-1
IUPAC NameN-methyl-N-(3-methylphenyl)-1-(naphthalen-2-yloxy)methanethioamide
Molecular FormulaC19H17NOS
InChI KeyFUSNMLFNXJSCDI-UHFFFAOYSA-N
SMILESCN(C(=S)OC1=CC=C2C=CC=CC2=C1)C1=CC=CC(C)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to yellow
Assay (unspecified)≥97.5%
FormPowder
Melting Point110-113°C
Tolnaftate is used to study the mechanism of ergosterol (sterol) biosynthesis at the rate-limiting step of squalene epoxidase inhibition. Tolnaftate is clinically used to treat cutaneous infections such as athlete?s foot, jock itch, and ringworm1. It also has been used as a comparator compound for antifungal efficiency studies.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in chloroform at 50 mg/ml. Sparingly soluble in ethanol or methanol

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature: 2 - 8°C
RUO – Research Use Only

General References:

  1. Craig Knox et. al. DrugBank 3.0: a comprehensive resource for omics research on drugs. Nucleic Acids Research. 2011, 39, D1035-D1041.
  2. J Pannu et. al. NB-002, a novel nanoemulsion with broad antifungal activity against dermatophytes, other filamentous fungi, and Candida albicans. Antimicrobial Agents and Chemotherapy. 2011, 53, (8), 3273-3279.