Thermo Scientific Chemicals

Tolnaftate, Thermo Scientific Chemicals

Catalog number: J61834.06
5 g, Each
Thermo Scientific Chemicals

Tolnaftate, Thermo Scientific Chemicals

Catalog number: J61834.06
5 g, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: J61834.06
also known as J61834-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
2398-96-1
IUPAC Name
N-methyl-N-(3-methylphenyl)-1-(naphthalen-2-yloxy)methanethioamide
Molecular Formula
C19H17NOS
InChI Key
FUSNMLFNXJSCDI-UHFFFAOYSA-N
SMILES
CN(C(=S)OC1=CC=C2C=CC=CC2=C1)C1=CC=CC(C)=C1
Appearance (Color)
White to yellow
Assay (unspecified)
≥97.5%
Form
Powder
Melting Point
110-113°C

Description

Tolnaftate is used to study the mechanism of ergosterol (sterol) biosynthesis at the rate-limiting step of squalene epoxidase inhibition. Tolnaftate is clinically used to treat cutaneous infections such as athlete?s foot, jock itch, and ringworm1. It also has been used as a comparator compound for antifungal efficiency studies.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in chloroform at 50 mg/ml. Sparingly soluble in ethanol or methanol

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature: 2 - 8°C
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text