Thermo Scientific Chemicals

Magnesium sulfate heptahydrate, Cell Culture Reagent, Thermo Scientific Chemicals

Catalog number: J61839.36
500 g, Each
Thermo Scientific Chemicals

Magnesium sulfate heptahydrate, Cell Culture Reagent, Thermo Scientific Chemicals

Catalog number: J61839.36
500 g, Each
Quantity
Catalog number: J61839.36
also known as J61839-36
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
10034-99-8
IUPAC Name
magnesium(2+) heptahydrate sulfate
Molecular Formula
H14MgO11S
InChI Key
WRUGWIBCXHJTDG-UHFFFAOYSA-L
SMILES
O.O.O.O.O.O.O.[Mg++].[O-]S([O-])(=O)=O
Form
Powder or crystals or crystalline powder
Appearance (Color)
Colorless to white
Assay from Supplier's CofA
≥99.0%

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text