Sodium pyruvate, Cell Culture Grade, Thermo Scientific Chemicals
Sodium pyruvate, Cell Culture Grade, Thermo Scientific Chemicals
Thermo Scientific™

Sodium pyruvate, Cell Culture Grade, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
100 g
Catalog number J61840.18
also known as J61840-18
Price (USD)/ Each
146.65
Online exclusive
163.00 
Save 16.35 (10%)
-
Add to cart
Quantity:
50 g
Request bulk or custom format
Price (USD)/ Each
146.65
Online exclusive
163.00 
Save 16.35 (10%)
Add to cart
Sodium pyruvate, Cell Culture Grade, Thermo Scientific Chemicals
Catalog numberJ61840.18
Price (USD)/ Each
146.65
Online exclusive
163.00 
Save 16.35 (10%)
-
Add to cart
Chemical Identifiers
CAS113-24-6
IUPAC Namesodium 2-oxopropanoate
Molecular FormulaC3H3NaO3
InChI KeyDAEPDZWVDSPTHF-UHFFFAOYSA-M
SMILES[Na+].CC(=O)C([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (Titration)≥99.0% (dried basis)
Loss on Drying≤2.0%
Heavy Metals≤10 ppm (as Pb)
pH5.0-7.0
Appearance (Color)White to cream
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only