Karaya Gum, Thermo Scientific Chemicals
Karaya Gum, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Karaya Gum, Thermo Scientific Chemicals

 
Have Questions?
Quantity:
500 g
Catalog number J61844.36
also known as J61844-36
Price (USD)/ Each
71.65
Online exclusive
79.00 
Save 7.35 (9%)
-
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (USD)/ Each
71.65
Online exclusive
79.00 
Save 7.35 (9%)
Add to cart
Karaya Gum, Thermo Scientific Chemicals
Catalog numberJ61844.36
Price (USD)/ Each
71.65
Online exclusive
79.00 
Save 7.35 (9%)
-
Add to cart
Chemical Identifiers
CAS9000-36-6
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)Identity: Conforms
Used as a thickener and emulsifier

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only