Thermo Scientific Chemicals

Papain, Carica Papaya Latex

Catalog number: J61875.MB
25 mg, Each
Thermo Scientific Chemicals

Papain, Carica Papaya Latex

Catalog number: J61875.MB
25 mg, Each
Quantity
Catalog number: J61875.MB
also known as J61875-MB
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
9001-73-4
Form
Lyophilized
units/mgP
Activates to at least 15.0 units per mg protein
Unit definition
One unit hydrolyzes one micromole of benzoyl-L-arginine ethyl ester per minute at 25º C, pH 6.2, after activation in a solution containing 1.1 mM EDTA, 0.067 mM beta merceptoethanol and 5.5 mM cystein

Description

Papain, from papaya latex, has been used to digest carotid artery smooth muscle cells. Papain is used in dissecting solutions. It is used to produce Fab fragments of antibodies. It is used for cell dissociation since it has been shown to be more effective and less damaging with certain tissues.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text