Orange G/blue loading dye (6X), Thermo Scientific Chemicals
Orange G/blue loading dye (6X), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Orange G/blue loading dye (6X), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 mL
10 mL
Catalog number J61877.AC
also known as J61877-AC
Price (USD)/ Each
160.00
-
Add to cart
Quantity:
25 mL
Request bulk or custom format
Price (USD)/ Each
160.00
Add to cart
Orange G/blue loading dye (6X), Thermo Scientific Chemicals
Catalog numberJ61877.AC
Price (USD)/ Each
160.00
-
Add to cart
Specifications
Chemical Name or MaterialOrange G/blue loading dye
Recommended StorageAmbient temperatures
TSCANo
Concentration6X
Solubility InformationNot miscible in water.
View more

Orange G/blue loading dye (6X) is used for loading DNA samples into gel electrophoresis wells and tracking migration during electrophoresis. It is used in used in molecular biology protocols. It is also used in nucleic acid gel electrophoresis & blotting. It is used to prepare DNA markers and samples for loading on agarose or polyacrylamide gels.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Orange G/blue loading dye (6X) is used for loading DNA samples into gel electrophoresis wells and tracking migration during electrophoresis. It is used in used in molecular biology protocols. It is also used in nucleic acid gel electrophoresis & blotting. It is used to prepare DNA markers and samples for loading on agarose or polyacrylamide gels.

Solubility
Not miscible in water.

Notes
Contains: 0.25% Orange-G, 0.25% bromophenol blue, 0.25% xylene cyanol FF, 15% Ficoll, 30mM EDTA in autoclaved DI water
RUO – Research Use Only

General References:

  1. RJ Williams. Isolation and characterization of an unknown restriction endonuclease. Nuclease Methods and Protocols. 2001, 160, (1), 431-442.
  2. PA Asare.; IKA Galyuon.; JK Sarfo.; JP Tetteh. Morphological and molecular based diversity studies of some cassava (Manihot esculenta crantz) germplasm in Ghana. African journal of biotechnology. 2013, 63, (10), 891-895.