Thermo Scientific Chemicals

Blasticidin S hydrochloride, ≥98%,/ml in 1M HEPES buffer soln., Thermo Scientific Chemicals

Catalog number: J61883.WDO
5 x 20 mg, Each
Thermo Scientific Chemicals

Blasticidin S hydrochloride, ≥98%,/ml in 1M HEPES buffer soln., Thermo Scientific Chemicals

Catalog number: J61883.WDO
5 x 20 mg, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: J61883.WDO
also known as J61883-W$
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
3513-03-9
IUPAC Name
6-(4-amino-2-oxo-1,2-dihydropyrimidin-1-yl)-3-[3-amino-5-(N-methylcarbamimidamido)pentanamido]-3,6-dihydro-2H-pyran-2-carboxylic acid hydrochloride
Molecular Formula
C17H27ClN8O5
InChI Key
YQXYQOXRCNEATG-UHFFFAOYNA-N
SMILES
Cl.CN(CCC(N)CC(=O)NC1C=CC(OC1C(O)=O)N1C=CC(N)=NC1=O)C(N)=N
Form
Liquid
Appearance (Color)
Clear
Assay from Supplier's CofA
≥98.0%
pH
7.2-7.5

Description

Blasticidin S hydrochloride is used to inhibit protein synthesis in prokaryotic and eukaryotic organisms. It also induces apoptosis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Blasticidin S hydrochloride is used to inhibit protein synthesis in prokaryotic and eukaryotic organisms. It also induces apoptosis.

Solubility
Soluble in water.

Notes
Store in cool place. Recommended to prepare and use solutions on the same day. Incompatible with strong oxidizing agents and strong bases.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text